missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 6000, Thermo Scientific Chemicals
$45.15 - $245.53
Chemical Identifiers
CAS | 9016-00-6 |
---|---|
Molecular Formula | (C2H6OSi)n |
Molecular Weight (g/mol) | 74.15 |
MDL Number | MFCD00084411 |
InChI Key | SEUDSDUUJXTXSV-UHFFFAOYSA-N |
IUPAC Name | dimethylsilanone |
SMILES | C[Si](C)(-*)O-* |
Description
Polydimethylsiloxane, trimethylsiloxy is used as reagents and Pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9016-00-6 | |
74.15 | |
SEUDSDUUJXTXSV-UHFFFAOYSA-N | |
C[Si](C)(-*)O-* |
(C2H6OSi)n | |
MFCD00084411 | |
dimethylsilanone |
Specifications
Colorless | |
9016-00-6 | |
<-40°C | |
MFCD00084411 | |
Soluble in aromatic hydrocarbon,partially soluble in aliphatic and insoluble in water. | |
SEUDSDUUJXTXSV-UHFFFAOYSA-N | |
dimethylsilanone | |
315°C (599°F) | |
1.4025 |
Viscous Liquid | |
>205°C | |
(C2H6OSi)n | |
0.950 g/mL | |
50 mL | |
C[Si](C)(-*)O-* | |
74.15 | |
Odorless | |
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 6,000 |
Safety and Handling
RTECSNumber : TQ2690000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only