Learn More
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 237, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 042496AD
Description
Polydimethylsiloxane, trimethylsiloxy is used in water-repellent, protective creams & lotions. It is also used as surfactant, is used in coating auxiliary agents. It is also used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Colorless | |
M.W. 237 | |
9016-00-6 | |
-86°C | |
MFCD00084411 | |
0.820 g/mL | |
Soluble in aromatic hydrocarbon,partially soluble in aliphatic and insoluble in water. | |
SEUDSDUUJXTXSV-UHFFFAOYSA-N | |
dimethylsilanone | |
39°C (102°F) | |
1.3825 |
Polydimethylsiloxane, trimethylsiloxy terminated | |
Liquid | |
152°C to 153°C | |
(C2H6OSi)n | |
UN1993 | |
Hexamethyldisiloxane polymers | |
50 mL | |
C[Si](C)(-*)O-* | |
74.15 | |
Odorless |
Chemical Identifiers
9016-00-6 | |
74.15 | |
SEUDSDUUJXTXSV-UHFFFAOYSA-N | |
dimethylsilanone |
(C2H6OSi)n | |
MFCD00084411 | |
Hexamethyldisiloxane polymers | |
C[Si](C)(-*)O-* |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
RTECSNumber : TQ2690000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only