missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 139,000

Manufacturer:  Alfa Aesar™ 44644A1

 View more versions of this product

Catalog No. AA44644A1



Specifications

Specifications

Polydimethylsiloxane, trimethylsiloxy terminated
9016-00-6
(CH3)3SiO(Si(CH3)2O)nSi(CH3)3
Not miscible with water. Insoluble in methanol,and ethylene glycol.
Odorless
>205°C
<-40°C
321°C (610°F)
M.W. 139,000
0.977 g/mL
MFCD00148360
Viscous Liquid
1.4035
Colorless
1kg

Chemical Identifiers

9016-00-6
(CH3)3SiO(Si(CH3)2O)nSi(CH3)3
MFCD00148360
Safety and Handling

Safety and Handling

RTECSNumber : TQ2690000

TSCA : No

Recommended Storage : Ambient temperatures

Documents