missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Polydimethylsiloxane, hydroxy terminated, M.W. 4200

Manufacturer:  Alfa Aesar™ 43769K2

 View more versions of this product

Catalog No. AA43769K2



Specifications

Specifications

Polydimethylsiloxane, hydroxy terminated
70131-67-8
HOSi
silanol, unii-079v3j9o3x, silanole, silanol terminated polydimethylsiloxane
O[Si]
45.092
Liquid
M.W. 4,200
0.97g/mL
MFCD01325012
XQSFXFQDJCDXDT-UHFFFAOYSA-N
hydroxysilicon
6327522
1L

Chemical Identifiers

70131-67-8
45.092
XQSFXFQDJCDXDT-UHFFFAOYSA-N
6327522
O[Si]
HOSi
MFCD01325012
silanol, unii-079v3j9o3x, silanole, silanol terminated polydimethylsiloxane
hydroxysilicon
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents