missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Polydimethylsiloxane, hydroxy terminated, M.W. 4200

Manufacturer:  Alfa Aesar™ 43769AD

 View more versions of this product

Catalog No. AA43769AD



Specifications

Specifications

Polydimethylsiloxane, hydroxy terminated
70131-67-8
HOSi
silanol
O[Si]
45.092
Liquid
M.W. 4,200
0.97g/mL
MFCD01325012
XQSFXFQDJCDXDT-UHFFFAOYSA-N
hydroxysilicon
6327522
50mL

Chemical Identifiers

70131-67-8
45.092
XQSFXFQDJCDXDT-UHFFFAOYSA-N
6327522
O[Si]
HOSi
MFCD01325012
silanol
hydroxysilicon
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents