missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PolyCo™ VR™ Sleeve Gloves

An ideal fusion of an 11mil nitrile glove and VR 4mil protective sleeve

Manufacturer:  PolyCo™ 41450

 View more versions of this product

Catalog No. 19-125-578



Description

Description

  • Flexible
  • High tactile sensitivity
  • Elastic cuffs to prevent slippage
  • Complete arm and hand protection
  • Overall length: 29 in.
  • Glove length: 12 in.
  • Latex-free
  • Made in USA
  • 50 Pairs per Case

Specifications

Specifications

Elastic
Glove: 12 in.; Overall: 29 in.
Medium
Glove: 11 mil; Sleeve: 4 mil
Blue
Nitrile
Yes
SDS
Documents