missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
PolyCo™ VR™ Sleeve Gloves
An ideal fusion of an 11mil nitrile glove and VR 4mil protective sleeve
Manufacturer: PolyCo™ 41550
Description
- Flexible
- High tactile sensitivity
- Elastic cuffs to prevent slippage
- Complete arm and hand protection
- Overall length: 29 in.
- Glove length: 12 in.
- Latex-free
- Made in USA
- 50 Pairs per Case
Specifications
Elastic | |
Glove: 12 in.; Overall: 29 in. | |
Large | |
Glove: 11 mil; Sleeve: 4 mil |
Blue | |
Nitrile | |
Yes |