missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PolyCo™ VR™ Sleeve Gloves DFS Item

An ideal fusion of an 11mil nitrile glove and VR 4mil protective sleeve

Supplier:  PolyCo™ 41350

 View more versions of this product

Catalog No. 19-125-577



Description

Description

  • Flexible
  • High tactile sensitivity
  • Elastic cuffs to prevent slippage
  • Complete arm and hand protection
  • Overall length: 29 in.
  • Glove length: 12 in.
  • Latex-free
  • Made in USA
  • 50 Pairs per Case

Specifications

Specifications

Elastic
Yes
12 in. (Glove), 29 in. (Overall)
SDS
Documents

Documents

Promotions

Promotions