PolyCo™ VR™ Protective Boot Covers
Provide knee-high, foot and lower leg protection in situations where biological and chemical liquids are present
Supplier: PolyCo™ 49550
Description
- Knee-high cover with elastic top
- Two supple weights provide flexible, lightweight coverage
- Available in different colors
- Standard or cleanroom packaged (for Class 100 controlled environments)
Specifications
Large | |
Blue | |
Heat sealed | |
Ideal for use where biological and chemical liquids are present |
Polyolefin | |
Elastic Top | |
4 mil |