missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Polycarbonate Erlenmeyer Flask with Dip Tube, Male Luer Lock, Sterile

Provide optimum visibility and durability

Manufacturer:  Corning™ C11465

 View more versions of this product

Catalog No. 15-100-153



Description

Description

Manufactured from optically clear polycarbonate for optimum visibility and durability.Corning™ Disposable Polycarbonate Erlenmeyer Flasks are ideal for suspension culture shake flask applications.
  • Can be used for cell culture or media storage
  • Graduated, wide mouth
  • Individually wrapped
  • Sterilized by gamma radiation and certified nonpyrogenic
  • RNase-/DNase-free
  • Aseptic Transfer cap for easy culture transfer
  • Flask Only is autoclavable according to furnished instructions
Specifications

Specifications

68 oz.
Polypropylene
With molded-in graduations
2L
Yes
Threaded
Orange
Aseptic Transfer Cap
48mm
Polycarbonate
0.2μm Vent, 1/4 in. dip tube, male luer lock disconnect
Gamma-irradiated
Documents