missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyaniline, emeraldine salt from p-toluenesulfonic acid

Manufacturer:  Alfa Aesar™ 4368309

 View more versions of this product

Catalog No. AA4368309



Specifications

Specifications

Polyaniline
25233-30-1
Powder
>300°C (decomposition)
Emeraldine salt from p-toluenesulfonic acid
MFCD00284320
Green
10g

Chemical Identifiers

25233-30-1
MFCD00284320
Documents