Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyaniline, emeraldine base

Conductive polymer

$104.11 - $104.11

Chemical Identifiers

Chemical Name or Material Polyaniline
CAS 25233-30-1
Name Note Emeraldine base
MDL Number MFCD00284320
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4258106

  SDS
alfa aesar
4258106
5g Each for $104.11
Description & Specifications

Specifications

Quantity 5g
Name Note Emeraldine base
Physical Form Powder
Chemical Name or Material Polyaniline
MDL Number MFCD00284320
CAS 25233-30-1