missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Poly(vinylidene fluoride)

Manufacturer:  Alfa Aesar™ 44080A3

 View more versions of this product

Catalog No. AA44080A3



Specifications

Specifications

Poly (vinylidene fluoride)
C2H2F2
1,1-difluoroethylene
BQCIDUSAKPWEOX-UHFFFAOYSA-N
1,1-difluoroethene
6369
Powder
White
2kg
24937-79-9
MFCD00084470
Soluble in organic esters,dimethylacetamide and amines. Insoluble in water.
C=C(F)F
64.035
CHEBI:82550
Odorless
155°C to 160°C

Chemical Identifiers

24937-79-9
64.035
BQCIDUSAKPWEOX-UHFFFAOYSA-N
6369
1,1-difluoroethene
C2H2F2
MFCD00084470
1,1-difluoroethylene
CHEBI:82550
C=C(F)F
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents