missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Poly(vinylidene chloride-co-acrylonitrile), 80/20

$54.30 - $204.95

Chemical Identifiers

CAS 9010-76-8
Molecular Formula (-CH2CCl2-)x[-CH2CH(CN)-]y
MDL Number MFCD00134022
Chemical Name or Material Poly (vinylidene chloride-co-acrylonitrile), 80/20
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4375722 SDS Alfa Aesar™
4375722
100g Each for $54.30
AA4375736 SDS Alfa Aesar™
4375736
500g Each for $204.95
Specifications Description & Specifications

Chemical Identifiers

9010-76-8
(-CH2CCl2-)x[-CH2CH(CN)-]y
MFCD00134022
Poly (vinylidene chloride-co-acrylonitrile), 80/20

Specifications

Powder
1.60g/mL
9010-76-8
(-CH2CCl2-)x[-CH2CH(CN)-]y
100g
MFCD00134022
Off-white
Poly (vinylidene chloride-co-acrylonitrile), 80/20

Safety and Handling

GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA