Please login to your online account to display your discounted pricing

Alfa Aesar™ Poly(vinyl formal)

$73.69 - $223.00

Chemical Identifiers

Chemical Name or Material Poly (vinyl formal)
CAS 9003-33-2
MDL Number MFCD00084463
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4419022

  SDS
alfa aesar™
4419022
100g Each for $73.69

AA4419036

  SDS
alfa aesar™
4419036
500g Each for $223.00

AA4419014

  SDS
alfa aesar™
4419014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Poly (vinyl formal)
CAS 9003-33-2
TSCA TSCA
Density 1.23g/mL
Physical Form Granular Powder
MDL Number MFCD00084463
Solubility Information Soluble in acetone, chloroform, dioxane, THF
Quantity 100g