missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Poly(vinyl formal)

Manufacturer:  Alfa Aesar™ 4419036

 View more versions of this product

Catalog No. AA4419036



Specifications

Specifications

Poly (vinyl formal)
1.23g/mL
MFCD00084463
Soluble in acetone, chloroform, dioxane, THF
C=COCOC=C
100.117
Granular Powder
9003-33-2
C5H8O2
1,1'-methylenebisoxy bisethene
DFFZYNIDEOOVAU-UHFFFAOYSA-N
ethenoxymethoxyethene
165717
500g

Chemical Identifiers

9003-33-2
100.117
DFFZYNIDEOOVAU-UHFFFAOYSA-N
165717
C=COCOC=C
C5H8O2
MFCD00084463
1,1'-methylenebisoxy bisethene
ethenoxymethoxyethene
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents