missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 85,000 to 120,000, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific AC450140250

 View more versions of this product

Catalog No. AC450140250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Poly(vinyl alcohol)
C2H4O
230.0°C to 240.0°C
IMROMDMJAWUWLK-UHFFFAOYSA-N
ethenol
11199
Solubility in water: soluble in hot water.
9002-89-5
elvanol, ethenol, homopolymer, gohsenol, hydroxyethene, hydroxyethylene, mowiol, polyvinyl alcohol, polyviol, poval, vinyl alcohol
25g
C=CO
44.053
Plastic bottle

Chemical Identifiers

9002-89-5
44.053
elvanol, ethenol, homopolymer, gohsenol, hydroxyethene, hydroxyethylene, mowiol, polyvinyl alcohol, polyviol, poval, vinyl alcohol
ethenol
C2H4O
IMROMDMJAWUWLK-UHFFFAOYSA-N
11199
C=CO
SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only