Please login to your online account to display your discounted pricing

Poly(vinyl alcohol), 99-100% hydrolyzed, approx. M.W. 86000, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  418120250

 View more versions of this product

Catalog No. AC418120250

  • / Each
 

Description & Specifications

Chemical Identifiers

Ash 1.2% max.
Infrared Spectrum Authentic
Loss on Drying 5% max.
Packaging Glass bottle
Viscosity 25 to 35mPa&middnot;s (20°C 4% aq.sol.)
Color Off-white to beige to golden
Quantity 25g
Chemical Name or Material Poly(vinyl alcohol)
Name Note 99-100% Hydrolyzed
CAS 9002-89-5
Physical Form Off-white to beige to golden powder
Assay 99 to 100 mole% (degree of hydrolysis)
Linear Formula (-CH2CH(OH)-)x
MDL Number MFCD00081922
Synonym PVA
RTECS Number TR8100000
TSCA TSCA

Specifications

Chemical Name or Material Poly(vinyl alcohol)
Name Note 99-100% Hydrolyzed
CAS 9002-89-5
Assay 99 to 100 mole% (degree of hydrolysis)
Linear Formula (-CH2CH(OH)-)x
MDL Number MFCD00081922
Synonym PVA

Indicator