Please login to your online account to display your discounted pricing

Poly(vinyl alcohol), 98% hydrolyzed, average M.W. 16000, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC183305000

 View more versions of this product

Catalog No. AC183305000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder or Granules
Quantity 500g
Color White to off-white
Assay 98 mole% min. (degree of hydrolysis)
Viscosity 6mPa&middnot;s (20°C 4% aq. solution)
Merck Index 15, 7706
Loss on Drying 5% max.
Linear Formula [-CH2CH(OH)-]n
Synonym PVA
Infrared Spectrum Authentic
Ash 1% max.
Chemical Name or Material Poly(vinyl alcohol)
MDL Number MFCD00081922
Packaging Glass bottle
CAS 9002-89-5

Specifications

Chemical Name or Material Poly(vinyl alcohol)
Assay 98 mole% min. (degree of hydrolysis)
Linear Formula [-CH2CH(OH)-]n
MDL Number MFCD00081922
Synonym PVA