Please login to your online account to display your discounted pricing

Poly(vinyl alcohol), 98% hydrolyzed, average M.W. 16000, ACROS Organics™

$44.28 - $146.53

Chemical Identifiers

Chemical Name or Material Poly(vinyl alcohol)
Assay 98 mole% min. (degree of hydrolysis)
Linear Formula [-CH2CH(OH)-]n
MDL Number MFCD00081922
Synonym PVA
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC183305000

  SDS
thermo fisher scientific
AC183305000
Glass bottle 500g Each for $146.53

AC183300250

  SDS
fisher scientific acros
183300250
Glass bottle 25g Each for $44.28
Description & Specifications

Specifications

Physical Form Powder or Granules
Quantity 500g
Color White to off-white
Assay 98 mole% min. (degree of hydrolysis)
Viscosity 6mPa&middnot;s (20°C 4% aq. solution)
Merck Index 15, 7706
Linear Formula [-CH2CH(OH)-]n
Synonym PVA
Loss on Drying 5% max.
Chemical Name or Material Poly(vinyl alcohol)
Ash 1% max.
MDL Number MFCD00081922
Infrared Spectrum Authentic
Packaging Glass bottle
CAS 9002-89-5

Indicator