Please login to your online account to display your discounted pricing

Poly(vinyl alcohol), 98% hydrolyzed, average M.W. 16000, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  183300250

 View more versions of this product

Catalog No. AC183300250

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder or Granules
Quantity 25g
Color White to off-white
Merck Index 15, 7706
Assay 98 mole% min. (degree of hydrolysis)
Viscosity 6mPa&middnot;s (20°C 4% aq. solution)
Ash 1% max.
Linear Formula [-CH2CH(OH)-]n
MDL Number MFCD00081922
CAS 9002-89-5
Synonym PVA
Loss on Drying 5% max.
Chemical Name or Material Poly(vinyl alcohol)
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material Poly(vinyl alcohol)
Assay 98 mole% min. (degree of hydrolysis)
Linear Formula [-CH2CH(OH)-]n
MDL Number MFCD00081922
Synonym PVA