Please login to your online account to display your discounted pricing

Poly(vinyl alcohol), 95% hydrolyzed, average M.W. 95000, ACROS Organics™


Quantity 1kg
Packaging Glass bottle

Manufacturer: fisher scientific acros  183290010

 View more versions of this product

Catalog No. AC183290010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Granular Powder
Quantity 1kg
Color White to cream
Merck Index 15, 7706
RTECS Number TR8100000
Assay 95.5 to 96.5 mole% (degree of hydrolysis)
TSCA TSCA
Viscosity 27 to 31 mPa&middnot;s (20°C, 4% aq. sol.)
Infrared Spectrum Authentic
Loss on Drying 5% max.
Packaging Glass bottle
Synonym PVA
Ash 0.9% max.
Linear Formula [-CH2CH(OH)-]n
CAS 9002-89-5
Name Note 95% Hydrolyzed, Average M.W. 95000
Chemical Name or Material Poly(vinyl alcohol)
MDL Number MFCD00081922

Specifications

Chemical Name or Material Poly(vinyl alcohol)
Name Note 95% Hydrolyzed, Average M.W. 95000
CAS 9002-89-5
Assay 95.5 to 96.5 mole% (degree of hydrolysis)
Linear Formula [-CH2CH(OH)-]n
MDL Number MFCD00081922
Synonym PVA