Please login to your online account to display your discounted pricing

Poly(vinyl alcohol), 95% hydrolyzed, average M.W. 95000, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Poly(vinyl alcohol)
Name Note 95% Hydrolyzed, Average M.W. 95000
CAS 9002-89-5
Assay 95.5 to 96.5 mole% (degree of hydrolysis)
Linear Formula [-CH2CH(OH)-]n
MDL Number MFCD00081922
Synonym PVA
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC183290010

  SDS
fisher scientific acros
183290010
Glass bottle 1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Granular Powder
Quantity 1kg
Color White to cream
Merck Index 15, 7706
RTECS Number TR8100000
TSCA TSCA
Assay 95.5 to 96.5 mole% (degree of hydrolysis)
Viscosity 27 to 31 mPa&middnot;s (20°C, 4% aq. sol.)
Infrared Spectrum Authentic
Loss on Drying 5% max.
Packaging Glass bottle
Chemical Name or Material Poly(vinyl alcohol)
Ash 0.9% max.
Name Note 95% Hydrolyzed, Average M.W. 95000
Linear Formula [-CH2CH(OH)-]n
MDL Number MFCD00081922
CAS 9002-89-5
Synonym PVA

Indicator