Please login to your online account to display your discounted pricing

Poly(vinyl alcohol), 78% hydrolyzed, ∽M.W. 6000, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Poly(vinyl alcohol)
Name Note 75% Hydrolyzed
CAS 9002-89-5
Assay 75 to 79 mole% (degree of hydrolysis)
Linear Formula [-CH2CH(OH)-]n
MDL Number MFCD00081922
Synonym PVA
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC302780250

  SDS
fisher scientific acros
302780250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder or Granules
Quantity 25g
Color White to straw
Assay 75 to 79 mole% (degree of hydrolysis)
TSCA TSCA
Viscosity 3.1mPa&middnot;s (20°C 4% aq. sol.)
RTECS Number TR8100000
Merck Index 15, 7706
Ash 1% max.
Infrared Spectrum Authentic
Loss on Drying 5% max.
Packaging Glass bottle
Volatiles ≤5%
Chemical Name or Material Poly(vinyl alcohol)
Name Note 75% Hydrolyzed
Linear Formula [-CH2CH(OH)-]n
MDL Number MFCD00081922
CAS 9002-89-5
Synonym PVA
Saponification Value 195 to 240mg KOH/g

Indicator