missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Poly(tetrafluoroethylene), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 178930050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
White | |
Powder | |
(C2F4)n | |
MFCD00084455 | |
PTFE | |
FC(F)(-*)C(F)(F)-* | |
Authentic | |
Glass Bottle |
Poly(tetrafluoroethylene) | |
9002-84-0 | |
(-CF2CF2-)n | |
2.0000g/mL | |
5 g | |
NaN | |
1% max. |
Chemical Identifiers
9002-84-0 | |
NaN | |
PTFE |
(C2F4)n | |
MFCD00084455 | |
FC(F)(-*)C(F)(F)-* |
Safety and Handling
RTECSNumber : KX4025000
TSCA : TSCA
RUO – Research Use Only