missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Poly(styrene sulfonic acid) sodium salt, M.W. 500,000, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04505022
Description
Poly(styrene sulfonic acid) sodium salt is used in superplastifier in cement, ion exchange resins, membranes in fuel cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Poly (styrene sulfonic acid) sodium salt | |
25704-18-1 | |
(C8H7NaO3S)n | |
100 g | |
Soluble in water and lower glycols. | |
[Na]OS(=O)(=O)C1=CC=C(C=C1)C(-*)C-* | |
184.21 | |
Powder |
M.W. 500,000 | |
Brown to White | |
MFCD00084449 | |
4-styrenesulfonic acid sodium salt | |
MAGFQRLKWCCTQJ-UHFFFAOYSA-N | |
4-ethenylbenzenesulfonic acid;molecular hydrogen;sodium | |
131675404 |
Chemical Identifiers
25704-18-1 | |
184.21 | |
MAGFQRLKWCCTQJ-UHFFFAOYSA-N | |
131675404 | |
[Na]OS(=O)(=O)C1=CC=C(C=C1)C(-*)C-* |
(C8H7NaO3S)n | |
MFCD00084449 | |
4-styrenesulfonic acid sodium salt | |
4-ethenylbenzenesulfonic acid;molecular hydrogen;sodium |
Safety and Handling
RTECSNumber : DB8550000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only