missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Poly(styrene-divinylbenzene), aminomethylated, 1% cross-linked, 200-400 mesh, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 4439406
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Poly (styrene-divinylbenzene) | |
Powder | |
C7H9N | |
benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine | |
WGQKYBSKWIADBV-UHFFFAOYSA-N | |
phenylmethanamine | |
7504 | |
200 to 400 Mesh |
Aminomethylated, 1% cross-linked | |
89551-24-6 | |
MFCD00146442 | |
5 g | |
C1=CC=C(C=C1)CN | |
107.156 | |
CHEBI:40538 |
Chemical Identifiers
89551-24-6 | |
107.156 | |
WGQKYBSKWIADBV-UHFFFAOYSA-N | |
7504 | |
phenylmethanamine |
C7H9N | |
MFCD00146442 | |
benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine | |
CHEBI:40538 | |
C1=CC=C(C=C1)CN |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures