Please login to your online account to display your discounted pricing

Alfa Aesar™ Poly(styrene-divinylbenzene), aminomethylated, 1% cross-linked, 200-400 mesh

$128.08 - $1,517.31

Chemical Identifiers

Chemical Name or Material Poly (styrene-divinylbenzene)
CAS 89551-24-6
Name Note Aminomethylated, 1% cross-linked
MDL Number MFCD00146442
Synonym Aminomethyl polystyrene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4439406

  SDS
alfa aesar™
4439406
5g Each for $128.08

AA4439414

  SDS
alfa aesar™
4439414
25g Each for $480.66

AA4439422

  SDS
alfa aesar™
4439422
100g Each for $1,517.31
Description & Specifications

Specifications

Chemical Name or Material Poly (styrene-divinylbenzene)
Name Note Aminomethylated, 1% cross-linked
Physical Form Powder
MDL Number MFCD00146442
CAS 89551-24-6
Synonym Aminomethyl polystyrene
Mesh Size 200 to 400 Mesh
Quantity 5g