missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Poly(styrene-divinylbenzene), aminomethylated, 1% cross-linked, 100-200 mesh

Formation of amide groups using Boc strategy, as a scavenger resin

Manufacturer:  Alfa Aesar™ 4439222

 View more versions of this product

Catalog No. AA4439222



Specifications

Specifications

Poly (styrene-divinylbenzene)
89551-24-6
MFCD00146442
Slightly soluble in water.
C1=CC=C(C=C1)CN
107.156
CHEBI:40538
100 to 200 Mesh
Aminomethylated, 1% cross-linked
C7H9N
1-phenylmethanamine
WGQKYBSKWIADBV-UHFFFAOYSA-N
phenylmethanamine
7504
Powder
100g

Chemical Identifiers

89551-24-6
107.156
WGQKYBSKWIADBV-UHFFFAOYSA-N
7504
phenylmethanamine
C7H9N
MFCD00146442
1-phenylmethanamine
CHEBI:40538
C1=CC=C(C=C1)CN
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents