missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Poly(styrene-divinylbenzene), 1% cross-linked, 100-200 mesh

Manufacturer:  Alfa Aesar™ 4329222

 View more versions of this product

Catalog No. AA4329222



Specifications

Specifications

Poly (styrene-divinylbenzene)
9003-70-7
MFCD00146442
CHRJZRDFSQHIFI-UHFFFAOYSA-N
1,2-bis(ethenyl)benzene;styrene
174664
100 to 200 Mesh
100g
1% cross-linked
C18H18
1,2-diethenylbenzene; styrene
C=CC1=CC=CC=C1.C=CC1=CC=CC=C1C=C
234.342
Beads
Odorless

Chemical Identifiers

9003-70-7
234.342
CHRJZRDFSQHIFI-UHFFFAOYSA-N
174664
C=CC1=CC=CC=C1.C=CC1=CC=CC=C1C=C
C18H18
MFCD00146442
1,2-diethenylbenzene; styrene
1,2-bis(ethenyl)benzene;styrene
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents