missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Poly(propylene glycol) bis(2-aminopropyl ether), Mn ca 230

Manufacturer:  Alfa Aesar™ H37543K4

 View more versions of this product

Catalog No. AAH37543K4


 

Specifications

Specifications

Poly(propylene glycol) bis(2-aminopropyl ether)
9046-10-0
UN2922
JCZPOYAMKJFOLA-UHFFFAOYSA-N
pyrrolidine-3,4-diol
13329881
0.948g/mL
2.5L
Mn ≈230
C4H9NO2
JEFFAMINE® D-230
C1C(C(CN1)O)O
103.121
Liquid
Colorless through Yellow

Chemical Identifiers

9046-10-0
103.121
JEFFAMINE® D-230
C4H9NO2
JCZPOYAMKJFOLA-UHFFFAOYSA-N
13329881
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

RTECSNumber : TR3702500

TSCA : TSCA

SDS Documents