missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Poly(propylene glycol), average M.W. 425, ACROS Organics™

Manufacturer:  Acros Organics 192150050

 View more versions of this product

Catalog No. AC192150050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Poly(propylene glycol)
25322-69-4
C6H14O3
MFCD00084446
CC(CO)OCC(C)O
134.175
Glass bottle
Solubility in water: completely soluble. Other solubilities: soluble in ethanol, acetone, benzene, toluene,, chloroform, dioxane, ether, thf
80 mm2/s (25°C)
-25.0°C
185°C
Average M.W. 425
1.0000g/mL
H[OCH(CH3)CH2]nOH
DUFKCOQISQKSAV-UHFFFAOYSA-N
2-(2-hydroxypropoxy)propan-1-ol
32881
1.4465 to 1.4505
1
200.0°C
5g

Chemical Identifiers

25322-69-4
134.175
DUFKCOQISQKSAV-UHFFFAOYSA-N
C6H14O3
MFCD00084446
32881
Safety and Handling

Safety and Handling

RTECSNumber : OE5784360

TSCA : TSCA

SDS Documents