missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Poly(propylene glycol), average M.W. 425, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 192155000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Poly(propylene glycol) | |
25322-69-4 | |
-25.0°C | |
H[OCH(CH3)CH2]nOH | |
1.0000g/mL | |
Solubility in water: completely soluble. Other solubilities: soluble in ethanol, acetone, benzene, toluene,, chloroform, dioxane, ether, thf | |
DNIAPMSPPWPWGF-UHFFFAOYNA-N | |
2-(2-hydroxypropoxy)propan-1-ol | |
32881 | |
Glass bottle | |
1 |
Average M.W. 425 | |
200.0°C | |
(C3H6O)n | |
MFCD00084446 | |
polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol | |
500 g | |
[H]OC(C)CO | |
134.18 | |
185°C | |
1.4465 to 1.4505 | |
80 mm2/s (25°C) |
Chemical Identifiers
25322-69-4 | |
134.18 | |
DNIAPMSPPWPWGF-UHFFFAOYNA-N | |
32881 | |
[H]OC(C)CO |
(C3H6O)n | |
MFCD00084446 | |
polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol | |
2-(2-hydroxypropoxy)propan-1-ol |
Safety and Handling
RTECSNumber : OE5784360
TSCA : TSCA
RUO – Research Use Only