missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Poly(propylene glycol), average M.W. 2.000, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 192180025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Poly(propylene glycol) | |
25322-69-4 | |
C6H14O3 | |
MFCD01779687 | |
polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol | |
DUFKCOQISQKSAV-UHFFFAOYSA-N | |
2-(2-hydroxypropoxy)propan-1-ol | |
32881 | |
53 to 58 mg KOH/g | |
Glass bottle | |
1 | |
0.15% Max. |
Average M.W. 2.000 | |
0.1 mg KOH/g max. | |
H[OCH(CH3)CH2]nOH | |
1.0000g/mL | |
2.5kg | |
CC(CO)OCC(C)O | |
134.175 | |
230°C | |
Authentic | |
1.4495 to 1.4515 | |
300 mm2/s (25°C) |
Safety and Handling
RTECSNumber : OE5784360
TSCA : TSCA
RUO â Research Use Only