Please login to your online account to display your discounted pricing

Poly(phenylene sulfide), powder, nominal M.W. 10000, ACROS Organics™


Quantity 250g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  178852500

 View more versions of this product

Catalog No. AC178852500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Quantity 250g
Color Off-white to tan
Density 1.3600g/mL
Chemical Name or Material Poly(phenylene sulfide)
Name Note nominal M.W. 10000
Linear Formula (-C6H4S-)n
MDL Number MFCD00084445
CAS 9016-75-5
Infrared Spectrum Authentic
Loss on Drying 1% max.
Packaging Plastic bottle

Specifications

Chemical Name or Material Poly(phenylene sulfide)
Name Note nominal M.W. 10000
CAS 9016-75-5
Linear Formula (-C6H4S-)n
MDL Number MFCD00084445