Please login to your online account to display your discounted pricing

Poly(phenylene sulfide), powder, nominal M.W. 10000, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Poly(phenylene sulfide)
Name Note nominal M.W. 10000
CAS 9016-75-5
Linear Formula (-C6H4S-)n
MDL Number MFCD00084445
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC178851000

  SDS
thermo fisher scientific
178851000
Plastic bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC178852500

  SDS
thermo fisher scientific
178852500
Plastic bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC178850050

  SDS
thermo fisher scientific
AC178850050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Quantity 100g
Color Off-white to tan
Density 1.3600g/mL
Infrared Spectrum Authentic
Chemical Name or Material Poly(phenylene sulfide)
Name Note nominal M.W. 10000
Linear Formula (-C6H4S-)n
MDL Number MFCD00084445
CAS 9016-75-5
Loss on Drying 1% max.
Packaging Plastic bottle