missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Poly[[N-isopropylacrylamide-co-[N-[3-(dodecyldimethylammonio)propyl]acrylamide]-co-(1,3,5-triacryloylhexahydro-1,3,5-triazine)]perruthenate], TCI America™

Manufacturer:  TCI America P16981G

 View more versions of this product

Catalog No. P16981G



Specifications

Specifications

Poly[[N-isopropylacrylamide-co-[N-[3-(dodecyldimethylammonio)propyl]acrylamide]-co-(1,3,5-triacryloylhexahydro-1,3,5-triazine)]perruthenate]
Black
1g
Crystal-Powder at 20°C

Chemical Identifiers

0.0
icon-pdf.png
Safety and Handling

Safety and Handling

TSCA : No

Documents