missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Poly(methyl methacrylate), beads, average M.W. 35,000, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 178760250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
White | |
nominal M.W. 38000 | |
9011-14-7 | |
(C5H8O2)n | |
MFCD00134349 | |
Methyl methacrylate, polymerized, PMMA | |
25 g | |
COC(=O)C(C)(-*)C-* | |
NaN | |
Authentic | |
Glass bottle | |
1.49 |
Poly(methyl methacrylate) | |
Beads | |
105.0°C | |
[-CH2C(CH3)(CO2CH3)-]n | |
1.18 | |
Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, chloroform, mek, thf | |
PMAMJWJDBDSDHV-UHFFFAOYSA-N | |
1-methoxy-2-methyl-1-oxopropan-2-yl | |
300°C | |
5% max. | |
Bead size (typically): 50 to 150 micron |
Chemical Identifiers
9011-14-7 | |
NaN | |
PMAMJWJDBDSDHV-UHFFFAOYSA-N | |
1-methoxy-2-methyl-1-oxopropan-2-yl |
(C5H8O2)n | |
MFCD00134349 | |
Methyl methacrylate, polymerized, PMMA | |
COC(=O)C(C)(-*)C-* |
Safety and Handling
RTECSNumber : TR0400000
TSCA : TSCA
RUO – Research Use Only