Please login to your online account to display your discounted pricing

Alfa Aesar™ Poly(methyl methacrylate)

$39.62 - $194.38

Chemical Identifiers

Chemical Name or Material Poly (methyl methacrylate)
CAS 9011-14-7
Molecular Formula [-CH2C(CH3)(CO2CH3)-]n
MDL Number MFCD00134349
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4398218

  SDS
alfa aesar™
4398218
50g Each for $39.62

AA4398230

  SDS
alfa aesar™
4398230
250g Each for $62.68

AA43982A1

  SDS
alfa aesar™
43982A1
1kg Each for $194.38
Description & Specifications

Specifications

Chemical Name or Material Poly (methyl methacrylate)
TSCA TSCA
Physical Form Powder
Molecular Formula [-CH2C(CH3)(CO2CH3)-]n
MDL Number MFCD00134349
CAS 9011-14-7
Color White
Melting Point >150°C
Quantity 50g
RTECS Number TR0400000