missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Poly(methyl methacrylate)

Manufacturer:  Alfa Aesar™ 4398230

 View more versions of this product

Catalog No. AA4398230



Specifications

Specifications

Poly (methyl methacrylate)
[-CH2C(CH3)(CO2CH3)-]n
Powder
>150°C
9011-14-7
MFCD00134349
White
250g

Chemical Identifiers

9011-14-7
[-CH2C(CH3)(CO2CH3)-]n
MFCD00134349
Safety and Handling

Safety and Handling

RTECSNumber : TR0400000

TSCA : TSCA

Documents