Please login to your online account to display your discounted pricing

Alfa Aesar™ Poly(methyl-3,3,3-trifluoropropylsiloxane), M.W. 4,600

Fluorosilicone lubricant

$27.08 - $253.99

Chemical Identifiers

Chemical Name or Material Poly (methyl-3,3,3-trifluoropropylsiloxane)
CAS 63148-56-1
Name Note M.W. 4,600
Molecular Formula [-Si(CH3)(CH3CH2CF3)O-]n
MDL Number MFCD00241477
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4322906

 
alfa aesar™
4322906
5g Each for $27.08

AA4322922

 
alfa aesar™
4322922
100g Each for $253.99
Description & Specifications

Specifications

Chemical Name or Material Poly (methyl-3,3,3-trifluoropropylsiloxane)
TSCA TSCA
Density 1.28g/mL
Name Note M.W. 4,600
Physical Form Liquid
Molecular Formula [-Si(CH3)(CH3CH2CF3)O-]n
MDL Number MFCD00241477
CAS 63148-56-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Refractive Index 1.382
Color Colorless
Quantity 5g
Flash Point 290°C (554°F)
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.