Learn More
Poly(methyl-3,3,3-trifluoropropylsiloxane), M.W. 14,000, Thermo Scientific Chemicals
Fluorosilicone lubricant
Supplier: Thermo Scientific Chemicals 04323022
Description
Fluorosilicone lubricant, lubricant additive, release agent, defoaming Agent, additive to improve chemical resistance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Poly (methyl-3,3,3-trifluoropropylsiloxane) | |
Liquid | |
C4H7F3OSi | |
1.30 g/mL | |
Insoluble in water. | |
CUMCLKHSZZGALD-UHFFFAOYSA-N | |
methyl-oxo-(3,3,3-trifluoropropyl)silane | |
4285173 | |
1.383 |
M.W. 14,000 | |
63148-56-1 | |
MFCD00241477 | |
methyl 3,3,3-trifluoropropyl silanone, poly methyl-3,3,3-trifluoropropylsiloxane, methyl-oxo-3,3,3-trifluoropropyl silane, 5,5,5-trifluoro-2-silapentane-2-one, poly 3,3,3-trifluoropropylmethylsiloxane, methyl 3,3,3-trifluoropropyl cyclopolysiloxanes, silane, methyloxo 3,3,3-trifluoropropyl, methyl 3,3,3-trifluoropropyl cyclopolysiloxanes, mixture of tri-and tetra | |
100 g | |
C[Si](=O)CCC(F)(F)F | |
156.179 | |
315°C (599°F) |
Chemical Identifiers
63148-56-1 | |
156.179 | |
CUMCLKHSZZGALD-UHFFFAOYSA-N | |
4285173 | |
C[Si](=O)CCC(F)(F)F |
C4H7F3OSi | |
MFCD00241477 | |
methyl 3,3,3-trifluoropropyl silanone, poly methyl-3,3,3-trifluoropropylsiloxane, methyl-oxo-3,3,3-trifluoropropyl silane, 5,5,5-trifluoro-2-silapentane-2-one, poly 3,3,3-trifluoropropylmethylsiloxane, methyl 3,3,3-trifluoropropyl cyclopolysiloxanes, silane, methyloxo 3,3,3-trifluoropropyl, methyl 3,3,3-trifluoropropyl cyclopolysiloxanes, mixture of tri-and tetra | |
methyl-oxo-(3,3,3-trifluoropropyl)silane |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only