Please login to your online account to display your discounted pricing

Poly-L-leucine, ACROS Organics™

$272.02 - $272.02

Chemical Identifiers

Chemical Name or Material Poly-L-leucine
CAS 25322-63-8
MDL Number MFCD00166370
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC327582500

  SDS
thermo fisher scientific
327582500
Glass bottle 250mg Each for $272.02
Description & Specifications

Specifications

Physical Form Powder
Quantity 250mg
Color White to off-white
Enantiomeric Excess 90% min. (chiral epoxidation of chalcone)
Packaging Glass bottle
CAS 25322-63-8
Chemical Name or Material Poly-L-leucine
MDL Number MFCD00166370