Please login to your online account to display your discounted pricing

Alfa Aesar™ Poly-L-leucine-1,3-diaminopropane


Quantity 250mg

Manufacturer: alfa aesar™  L19272MD

 View more versions of this product

Catalog No. AAL19272MD

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Poly-L-leucine-1,3-diaminopropane
Quantity 250mg
MDL Number MFCD03100958

Specifications

Chemical Name or Material Poly-L-leucine-1,3-diaminopropane
MDL Number MFCD03100958