Please login to your online account to display your discounted pricing

Alfa Aesar™ Poly-L-leucine-1,3-diaminopropane

$224.73 - $224.73

Chemical Identifiers

Chemical Name or Material Poly-L-leucine-1,3-diaminopropane
MDL Number MFCD03100958
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1927203

 
alfa aesar™
L1927203
1g Each for $224.73

AAL19272MD

 
alfa aesar™
L19272MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

MDL Number MFCD03100958
Chemical Name or Material Poly-L-leucine-1,3-diaminopropane
Quantity 1g