missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Poly(ethylene glycol methyleether), average M.W. 350, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 192305000

 View more versions of this product

Catalog No. AC192305000



Specifications

Specifications

Poly(ethylene glycol methyleether)
9004-74-4
C3H8O2
MFCD00084416
XNWFRZJHXBZDAG-UHFFFAOYSA-N
152 to 168mg KOH/g
76.095
CHEBI:46790
Liquid
1.4560 to 1.4580 (20°C, 589nm)
Undesignated
500g
Average M.W. 350
1.09g/mL
CH3(OCH2CH)nOH
2-methoxy-1-ethanol
COCCO
2-methoxyethanol
8019
Authentic
Glass bottle
0.15% max. (K.F.)
4.0 to 8.0 (1% aq. soln.)

Chemical Identifiers

9004-74-4
76.095
XNWFRZJHXBZDAG-UHFFFAOYSA-N
8019
2-methoxyethanol
C3H8O2
MFCD00084416
2-methoxy-1-ethanol
CHEBI:46790
COCCO
Safety and Handling

Safety and Handling

RTECSNumber : PE8263000

TSCA : TSCA

SDS Documents