missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Poly(ethylene glycol methyleether), average M.W. 350, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 192300010

 View more versions of this product

Catalog No. AC192300010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Liquid
Clear
1.09g/mL
0.15% max. (K.F.)
Glass bottle
Average M.W. 350
Poly(ethylene glycol methyleether)
9004-74-4
1kg
4.0 to 8.0 (1% aq. soln.)
152 to 168mg KOH/g
Authentic
1.4560 to 1.4580 (20°C, 589nm)
CH3(OCH2CH)nOH
MFCD00084416

Chemical Identifiers

9004-74-4
CH3(OCH2CH)nOH
MFCD00084416
Poly(ethylene glycol methyleether), Average M.W. 350

Safety and Handling

SDS & Safety Documents

RTECSNumber : PE8263000

TSCA : TSCA