missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Poly(ethylene glycol), average M.W. 4000, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC434630010
Specifications
Poly(ethylene glycol) | |
Plastic Bottle | |
White | |
1kg | |
H(OCH2CH2)nOH | |
PEO | |
4000 |
Authentic | |
0.5% max. | |
54.0°C to 58.0°C | |
25322-68-3 | |
15,7688 | |
4000 | |
Flakes |
Chemical Identifiers
25322-68-3 | |
4000 | |
PEO |
Safety and Handling
EINECSNumber : 500-038-2