missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Poly(ethylene glycol), average M.W. 300, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 192220025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Clear liquid | |
25322-68-3 | |
(C2H4O)n | |
MFCD01779601 | |
15, 7688 | |
Solubility in water: soluble. Other solubilities: soluble in many organic solvents | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol | |
>180°C | |
Glass bottle | |
1.12 |
Poly(ethylene glycol) | |
-15.0°C to +10.0°C | |
H(OCH2CH2)nOH | |
1.1200g/mL | |
PEG | |
2.5 L | |
[H]OCCO | |
62.07 | |
356 to 394mg KOH/g | |
1.4620 to 1.4650 | |
85-100 mPa.s (20°C) |
Chemical Identifiers
25322-68-3 | |
62.07 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol |
(C2H4O)n | |
MFCD01779601 | |
PEG | |
[H]OCCO |
Safety and Handling
EINECSNumber : 500-038-2
RUO – Research Use Only