missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Poly(ethylene glycol), average M.W. 200, Thermo Scientific Chemicals
$32.06 - $122.40
Chemical Identifiers
CAS | 25322-68-3 |
---|---|
Molecular Formula | (C2H4O)n |
Molecular Weight (g/mol) | 62.07 |
MDL Number | MFCD01779601 |
InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
Synonym | PEG |
IUPAC Name | ethane-1,2-diol |
SMILES | [H]OCCO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC192210050
|
Thermo Scientific Chemicals
192210050 |
5 mL | Plastic bottle |
Each for $32.06
|
|
||||
AC192210010
|
Thermo Scientific Chemicals
192210010 |
1 L | Plastic bottle |
Each for $122.40
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
25322-68-3 | |
62.07 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol |
(C2H4O)n | |
MFCD01779601 | |
PEG | |
[H]OCCO |
Specifications
Liquid | |
-65.0°C | |
H(OCH2CH2)nOH | |
1.1250g/mL | |
Solubility in water: soluble. Other solubilities: soluble in many organic solvents | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol | |
200 | |
534 to 591mg KOH/g | |
Plastic bottle | |
1.125 | |
0.5% max. (K.F.) |
25322-68-3 | |
(C2H4O)n | |
MFCD01779601 | |
PEG | |
5 mL | |
[H]OCCO | |
62.07 | |
171°C | |
Authentic | |
1.4580 to 1.4610 | |
60-70 mPa.s (20°C) | |
Poly(ethylene glycol) |
RUO – Research Use Only